siRNA Details
Gene Symbol: MTFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016498.4 | NP_057582.2 | 3 | 516 | ||
GenBank mRNA | AF151076.1 | - | 3 | 478 | ||
AK026104.1 | - | 3 | 384 | |||
BC001608.1 | - | 3 | 342 | 3542567 | ||
BC009300.2 | - | 3 | 374 | 4126237 | ||
BC014446.1 | - | 3 | 342 | 4855192 | ||
BC030989.2 | - | 3 | 403 | 4077757 | ||
BC038831.2 | - | 3 | 407 | 3908299 | ||
BC046132.1 | - | 3 | 306 | 4446034 | ||
CR456345.1 | - | 3 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
