siRNA Details
Gene Symbol: TMEM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018126.2 | NP_060596.2 | 6 | 911 | ||
XM_005248116.3 | XP_005248173.1 | 7 | 750 | |||
XM_005248117.2 | XP_005248174.1 | 7 | 698 | |||
GenBank mRNA | AK001387.1 | - | 6 | 879 | ||
AK023062.1 | - | 6 | 891 | |||
AK094190.1 | - | 7 | 822 | |||
AK308501.1 | - | 5 | 691 | |||
AY659966.1 | - | 6 | 879 | |||
BC000948.2 | - | 6 | 641 | 3448888 | ||
CR457260.1 | - | 6 | 546 |
