siRNA Details
Gene Symbol: NIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020202.4 | NP_064587.1 | 7, 8 | 663 | ||
GenBank mRNA | AF260334.1 | - | 7, 8 | 662 | ||
AF284574.1 | - | 7, 8 | 610 | |||
AK225979.1 | - | 6, 7 | 1141 | |||
AK313704.1 | - | 7, 8 | 605 | |||
BC020620.1 | - | 7, 8 | 620 | 4775062 | ||
BC107890.1 | - | 7, 8 | 651 | 6648910 | ||
FJ224326.1 | - | 7, 8 | 610 |
