siRNA Details
Gene Symbol: RTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ251832.1 | - | 2 | 565 | ||
AY562238.1 | - | 2 | 511 | |||
BC013161.1 | - | 2 | 573 | 3919961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
