siRNA Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170635.1 | NP_001164106.1 | 6, 7 | 962 | ||
NM_022737.2 | NP_073574.2 | 6, 7 | 1037 | |||
XM_005260017.1 | XP_005260074.1 | 6, 7 | 1037 | |||
XM_017027147.1 | XP_016882636.1 | 6, 7 | 1040 | |||
XM_017027148.1 | XP_016882637.1 | 6, 7 | 958 | |||
GenBank mRNA | AK023117.1 | - | 6, 7 | 998 | ||
AK094869.1 | - | 6, 7 | 850 | |||
AK225482.1 | - | 6, 7 | 952 | |||
AY304516.1 | - | 6, 7 | 992 | |||
AY339628.1 | - | 5, 6 | 692 | |||
BC009378.1 | - | 6, 7 | 880 | 4125316 |
