siRNA Details
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 21 | 2714 | ||
XM_005269357.3 | XP_005269414.1 | 21 | 2735 | |||
XM_011513120.2 | XP_011511422.1 | 22 | 2836 | |||
XM_011513121.1 | XP_011511423.1 | 21 | 2714 | |||
XM_011513122.1 | XP_011511424.1 | 21 | 2408 | |||
XM_011513123.2 | XP_011511425.1 | 22 | 2841 | |||
XM_011513124.2 | XP_011511426.1 | 22 | 2841 | |||
GenBank mRNA | AJ306929.1 | - | 12 | 3238 | ||
AK024639.1 | - | 10 | 1111 | |||
AK075094.1 | - | 2 | 159 | |||
AK225064.1 | - | 10 | 1111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
