siRNA Details
Gene Symbol: SPAG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024532.4 | NP_078808.3 | 16 | 1936 | ||
NR_047659.1 | 18 | 2111 | ||||
NR_047660.1 | 15 | 1817 | ||||
XM_011511814.1 | XP_011510116.1 | 15 | 1984 | |||
XM_011511819.2 | XP_011510121.1 | 15 | 2635 | |||
XM_011511820.1 | XP_011510122.1 | 15 | 1701 | |||
XM_011511821.2 | XP_011510123.1 | 15 | 2761 | |||
XM_011511822.1 | XP_011510124.1 | 14 | 1617 | |||
XM_011511823.2 | XP_011510125.1 | 14 | 2539 | |||
XM_011511835.2 | XP_011510137.1 | 14 | 1869 | |||
XM_017004896.1 | XP_016860385.1 | 17 | 2016 | |||
XM_017004897.1 | XP_016860386.1 | 14 | 4633 | |||
XM_017004898.1 | XP_016860387.1 | 11 | 1365 | |||
XM_017004900.1 | XP_016860389.1 | 15 | 1768 | |||
XM_017004901.1 | XP_016860390.1 | 15 | 1750 | |||
XM_017004902.1 | XP_016860391.1 | 9 | 1086 | |||
GenBank mRNA | AF310672.1 | - | 16 | 1873 | ||
AF490390.1 | - | 16 | 1866 | |||
AK302351.1 | - | 15 | 1759 | |||
BC025379.1 | - | 18 | 2116 | 4820489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
