siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024616.2 | NP_078892.2 | 3, 4 | 470 | ||
XM_011513133.2 | XP_011511435.1 | 3, 4 | 379 | |||
XM_017007181.1 | XP_016862670.1 | 3, 4 | 1444 | |||
GenBank mRNA | AK026839.1 | - | 3, 4 | 414 | ||
AY830714.1 | - | 3, 4 | 416 | |||
BC017064.1 | - | 3, 4 | 403 | 3885219 |
