siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171747.1 | NP_001165218.1 | 4 | 1157 | ||
NM_024616.2 | NP_078892.2 | 6 | 1410 | |||
XM_011513133.2 | XP_011511435.1 | 6 | 1319 | |||
XM_017007181.1 | XP_016862670.1 | 6 | 2384 | |||
GenBank mRNA | AA215562.1 | - | 1 | 244 | ||
AK026839.1 | - | 6 | 1354 | |||
AY830714.1 | - | 6 | 1356 | |||
BC017064.1 | - | 6 | 1343 | 3885219 |
