siRNA Details
Gene Symbol: ZNF385D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024697.2 | NP_078973.1 | 5 | 913 | ||
XM_011534122.1 | XP_011532424.1 | 7 | 1012 | |||
XM_017007191.1 | XP_016862680.1 | 7 | 951 | |||
XM_017007192.1 | XP_016862681.1 | 6 | 894 | |||
XM_017007193.1 | XP_016862682.1 | 7 | 941 | |||
XM_017007194.1 | XP_016862683.1 | 8 | 1006 | |||
XM_017007195.1 | XP_016862684.1 | 6 | 646 | |||
XM_017007196.1 | XP_016862685.1 | 6 | 1124 | |||
XM_017007197.1 | XP_016862686.1 | 6 | 1362 | |||
XM_017007198.1 | XP_016862687.1 | 6 | 1117 | |||
XM_017007199.1 | XP_016862688.1 | 5 | 472 | |||
XM_017007200.1 | XP_016862689.1 | 5 | 2688 | |||
XM_017007201.1 | XP_016862690.1 | 5 | 493 | |||
XM_017007202.1 | XP_016862691.1 | 3 | 209 | |||
GenBank mRNA | AK026072.1 | - | 5 | 913 | ||
AK310298.1 | - | 7 | 924 | |||
BC007212.1 | - | 5 | 737 | 3347783 |
