siRNA Details
Gene Symbol: CEBPA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026887.1 | 1 | 1219 | |||
GenBank mRNA | AK022417.1 | - | 1 | 1121 | ||
BC027959.1 | - | 1 | 1202 | |||
BC113872.1 | - | 1 | 1006 | |||
BC114482.1 | - | 1 | 1006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
