siRNA Details
Gene Symbol: TMEM191A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026815.1 | 1 | 375 | |||
GenBank mRNA | AK313335.1 | - | 1 | 556 | ||
AL136879.1 | - | 1 | 375 | |||
BC126323.1 | - | 1 | 342 | |||
BC126325.1 | - | 1 | 342 | |||
CR456359.1 | - | 1 | 360 |
Gene Symbol: LOC100996401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003846301.4 | XP_003846349.1 | 1 | 406 |
