siRNA Details
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032285.2 | NP_115661.1 | 3 | 458 | ||
XM_011528355.2 | XP_011526657.1 | 3 | 480 | |||
XM_011528357.2 | XP_011526659.1 | 4 | 665 | |||
GenBank mRNA | AL834276.1 | - | 3 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
