siRNA Details
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099737.2 | NP_001093207.1 | 6 | 694 | ||
NM_032332.3 | NP_115708.1 | 4 | 437 | |||
NR_029375.1 | 6 | 677 | ||||
GenBank mRNA | AK074525.1 | - | 6 | 667 | ||
AK223195.1 | - | 4 | 387 | |||
AK314164.1 | - | 6 | 671 | |||
BC005870.2 | - | 4 | 370 | 2961616 | ||
CB143206.1 | - | 4 | 437 |
