siRNA Details
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003058.3 | NP_003049.2 | 3 | 813 | ||
GenBank mRNA | AB075951.1 | - | 3 | 817 | ||
AK290787.1 | - | 3 | 717 | |||
BC030978.1 | - | 3 | 813 | |||
BC039899.1 | - | 3 | 694 | 5186229 | ||
X98333.1 | - | 3 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
