siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032433.2 | NP_115809.1 | 12 | 1765 | ||
XM_005260098.3 | XP_005260155.1 | 13 | 2078 | |||
XM_006722924.2 | XP_006722987.1 | 12 | 1842 | |||
XM_011528362.2 | XP_011526664.1 | 12 | 1806 | |||
XM_011528363.2 | XP_011526665.1 | 14 | 2906 | |||
XM_011528365.2 | XP_011526667.1 | 14 | 2247 | |||
XM_011528366.2 | XP_011526668.1 | 13 | 2075 | |||
XM_011528367.2 | XP_011526669.1 | 13 | 2088 | |||
XM_011528368.2 | XP_011526670.1 | 13 | 1995 | |||
XM_011528369.2 | XP_011526671.1 | 13 | 2008 | |||
XM_011528371.2 | XP_011526673.1 | 6 | 2172 | |||
XM_011528372.2 | XP_011526674.1 | 5 | 1101 | |||
XM_017027367.1 | XP_016882856.1 | 12 | 1782 | |||
XM_017027368.1 | XP_016882857.1 | 11 | 1650 | |||
XM_017027369.1 | XP_016882858.1 | 13 | 3403 | |||
XM_017027370.1 | XP_016882859.1 | 13 | 3416 | |||
XM_017027371.1 | XP_016882860.1 | 13 | 2028 | |||
GenBank mRNA | AB058709.1 | - | 5 | 1079 | ||
AF372702.1 | - | 12 | 1722 | |||
AK094028.1 | - | 13 | 2008 | |||
AK095142.1 | - | 1 | 647 | |||
AK122817.1 | - | 5 | 1646 | |||
AL832544.1 | - | 6 | 1257 | |||
BX647475.1 | - | 12 | 1726 |
