siRNA Details
Gene Symbol: KBTBD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032505.2 | NP_115894.2 | 2, 3 | 276 | ||
GenBank mRNA | AF385438.1 | - | 1, 2 | 209 | ||
AK096640.1 | - | 2, 3 | 272 | |||
BC117487.1 | - | 1, 2 | 173 | 40126038 | ||
DA246470.1 | - | 2, 3 | 272 | |||
DB065239.1 | - | 2, 3 | 277 |
