siRNA Details
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 8 | 6612 | ||
NM_005036.4 | NP_005027.2 | 9 | 6696 | |||
GenBank mRNA | AB073605.1 | - | 1 | 589 | ||
AK091885.1 | - | 1 | 401 | |||
BC004162.2 | - | 1 | 2755 | |||
BC071932.1 | - | 1 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
