siRNA Details
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 4 | 461 | ||
XM_005265537.4 | XP_005265594.1 | 4 | 474 | |||
XM_006713384.3 | XP_006713447.1 | 4 | 472 | |||
XM_011534214.2 | XP_011532516.1 | 4 | 476 | |||
XM_011534215.2 | XP_011532517.1 | 4 | 476 | |||
XM_011534216.2 | XP_011532518.1 | 3 | 376 | |||
XM_017007460.1 | XP_016862949.1 | 4 | 473 | |||
XM_017007461.1 | XP_016862950.1 | 4 | 462 | |||
GenBank mRNA | AJ318099.1 | - | 4 | 445 | ||
BC043595.1 | - | 4 | 454 | 5769824 |
