siRNA Details
Gene Symbol: PCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026555.1 | 2 | 470 | |||
GenBank mRNA | AF388200.1 | - | 2 | 465 | ||
AF462307.1 | - | 2 | 420 | |||
AK056825.1 | - | 2 | 470 | |||
BC105591.1 | - | 2 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
