siRNA Details
Gene Symbol: DOK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018110.3 | NP_060580.2 | 5 | 661 | ||
XM_005256042.3 | XP_005256099.1 | 4 | 603 | |||
XM_005256043.2 | XP_005256100.1 | 4 | 603 | |||
GenBank mRNA | AF466369.1 | - | 4 | 311 | ||
AK001350.1 | - | 5 | 588 | |||
AK307270.1 | - | 3 | 445 | |||
AK315426.1 | - | 4 | 571 | |||
BC001540.2 | - | 4 | 360 | 2988350 | ||
BC003541.1 | - | 5 | 603 | 3611472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
