siRNA Details
Gene Symbol: NCBP2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024388.1 | 1 | 770 | |||
GenBank mRNA | AK123020.1 | - | 1 | 721 | ||
AL832915.1 | - | 1 | 708 | |||
BC000257.1 | - | 1 | 704 | |||
BC007882.2 | - | 1 | 683 | |||
BC062368.1 | - | 1 | 770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
