siRNA Details
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301684.1 | NP_001288613.1 | 7 | 993 | ||
NM_138803.3 | NP_620158.3 | 9 | 1249 | |||
XM_005246319.3 | XP_005246376.1 | 10 | 1136 | |||
XM_011510638.2 | XP_011508940.1 | 10 | 1137 | |||
XM_017003392.1 | XP_016858881.1 | 9 | 1249 | |||
XM_017003393.1 | XP_016858882.1 | 9 | 932 | |||
XM_017003394.1 | XP_016858883.1 | 8 | 1082 | |||
XM_017003395.1 | XP_016858884.1 | 8 | 1145 | |||
XM_017003396.1 | XP_016858885.1 | 8 | 1145 | |||
XM_017003397.1 | XP_016858886.1 | 6 | 622 | |||
XM_017003398.1 | XP_016858887.1 | 3 | 578 | |||
GenBank mRNA | AF458588.1 | - | 9 | 1198 | ||
AK123220.1 | - | 4 | 478 | |||
AK308783.1 | - | 7 | 993 | |||
AK309246.1 | - | 8 | 1082 | |||
AK311042.1 | - | 7 | 1023 | |||
BC105928.1 | - | 7 | 806 | |||
BC107596.1 | - | 7 | 806 | 40025915 | ||
BC107597.1 | - | 9 | 1062 | 40025920 |
