siRNA Details
Gene Symbol: LOC105379554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546267.2 | XP_011544569.1 | 1 | 294 |
Gene Symbol: LOC100233156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037871.1 | 5 | 584 | |||
NR_037872.1 | 4 | 1438 | ||||
GenBank mRNA | AK092792.1 | - | 4 | 1419 | ||
BC104658.1 | - | 5 | 551 |
Gene Symbol: MAFIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096952.1 | - | 8 | 2455 | ||
DQ786282.1 | - | 5 | 637 |
Gene Symbol: TEKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286559.1 | NP_001273488.1 | 6 | 2034 | ||
NM_144705.3 | NP_653306.1 | 6 | 1331 | |||
XM_005263876.2 | XP_005263933.1 | 5 | 4842 | |||
XM_011510670.1 | XP_011508972.1 | 6 | 5120 | |||
GenBank mRNA | AK097438.1 | - | 6 | 2034 | ||
BC021716.2 | - | 6 | 1277 | 4831157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
