siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 40, 41 | 6757 | ||
XM_006711956.2 | XP_006712019.1 | 40, 41 | 6862 | |||
XM_011532649.2 | XP_011530951.1 | 40, 41 | 6711 | |||
XM_011532650.2 | XP_011530952.1 | 40, 41 | 6712 | |||
XM_011532652.1 | XP_011530954.1 | 39, 40 | 6631 | |||
XM_011532653.1 | XP_011530955.1 | 40, 41 | 6757 | |||
XM_011532654.1 | XP_011530956.1 | 39, 40 | 6490 | |||
XM_011532655.2 | XP_011530957.1 | 36, 37 | 5779 | |||
XM_011532657.1 | XP_011530959.1 | 34, 35 | 5518 | |||
XM_011532660.1 | XP_011530962.1 | 40, 41 | 6757 | |||
XM_017003521.1 | XP_016859010.1 | 39, 40 | 6631 | |||
XM_017003522.1 | XP_016859011.1 | 35, 36 | 6583 | |||
XM_017003523.1 | XP_016859012.1 | 35, 36 | 5750 | |||
GenBank mRNA | AB051484.2 | - | 4, 5 | 692 | ||
AB290165.1 | - | 6, 7 | 896 | |||
BC015442.1 | - | 5, 6 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
