siRNA Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148894.2 | NP_683692.2 | 2 | 392 | ||
XM_005248150.3 | XP_005248207.1 | 2 | 409 | |||
XM_005248151.3 | XP_005248208.1 | 2 | 409 | |||
XM_006713958.3 | XP_006714021.1 | 2 | 409 | |||
XM_011513827.2 | XP_011512129.1 | 2 | 409 | |||
XM_011513829.2 | XP_011512131.1 | 2 | 409 | |||
XM_017008009.1 | XP_016863498.1 | 2 | 409 | |||
XM_017008010.1 | XP_016863499.1 | 2 | 409 | |||
GenBank mRNA | AF528529.1 | - | 2 | 392 | ||
BC065546.1 | - | 2 | 341 | |||
BC087835.1 | - | 2 | 398 |
