siRNA Details
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007099.2 | NP_001007100.1 | 5 | 867 | ||
NM_001007100.2 | NP_001007101.1 | 5 | 858 | |||
NM_001007250.2 | NP_001007251.1 | 4 | 737 | |||
NM_001193599.1 | NP_001180528.1 | 15 | 2092 | |||
NM_001193600.1 | NP_001180529.1 | 15 | 2032 | |||
NM_001193617.1 | NP_001180546.1 | 15 | 2106 | |||
NM_002979.4 | NP_002970.2 | 16 | 2164 | |||
XM_005271103.4 | XP_005271160.1 | 15 | 1977 | |||
XM_017002046.1 | XP_016857535.1 | 6 | 1432 | |||
GenBank mRNA | AB208789.1 | - | 17 | 4859 | ||
AK294631.1 | - | 15 | 2035 | |||
BC005911.1 | - | 5 | 857 | |||
BX537618.1 | - | 3 | 3373 | |||
BX537619.1 | - | 3 | 3373 | |||
M55421.1 | - | 5 | 867 | |||
M75883.1 | - | 16 | 2017 | |||
M75884.1 | - | 8 | 1303 | |||
S52450.1 | - | 5 | 867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
