siRNA Details
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 11 | 2129 | ||
XM_005247950.3 | XP_005248007.1 | 11 | 2126 | |||
XM_011513407.2 | XP_011511709.1 | 11 | 2400 | |||
XM_011513408.1 | XP_011511710.1 | 12 | 2270 | |||
XM_011513409.2 | XP_011511711.1 | 12 | 2241 | |||
GenBank mRNA | AK092387.1 | - | 2 | 196 | ||
AK093044.1 | - | 7 | 1135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
