siRNA Details
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 11 | 2573 | ||
XM_005247950.3 | XP_005248007.1 | 11 | 2570 | |||
XM_011513407.2 | XP_011511709.1 | 11 | 2844 | |||
XM_011513408.1 | XP_011511710.1 | 12 | 2714 | |||
XM_011513409.2 | XP_011511711.1 | 12 | 2685 | |||
GenBank mRNA | AK092387.1 | - | 2 | 640 | ||
AK093044.1 | - | 7 | 1579 | |||
AK096417.1 | - | 1 | 21 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
