siRNA Details
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054875.1 | - | 3 | 1448 | ||
AL832097.1 | - | 1 | 152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C20orf166-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054875.1 | - | 3 | 1448 | ||
AL832097.1 | - | 1 | 152 |