siRNA Details
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 24 | 2915 | ||
XM_006712438.3 | XP_006712501.1 | 23 | 2852 | |||
XM_006712439.3 | XP_006712502.1 | 8 | 1499 | |||
XM_011510985.2 | XP_011509287.1 | 22 | 2644 | |||
XM_011510987.2 | XP_011509289.1 | 16 | 1835 | |||
XM_011510988.2 | XP_011509290.1 | 20 | 2361 | |||
XM_017003861.1 | XP_016859350.1 | 22 | 2759 | |||
XM_017003862.1 | XP_016859351.1 | 22 | 2738 | |||
XM_017003863.1 | XP_016859352.1 | 22 | 2657 | |||
XM_017003865.1 | XP_016859354.1 | 20 | 2376 | |||
XM_017003866.1 | XP_016859355.1 | 17 | 2051 | |||
GenBank mRNA | AK001064.1 | - | 1 | 1044 | ||
AK125768.1 | - | 8 | 1883 | |||
AK302685.1 | - | 19 | 2283 | |||
AL834537.1 | - | 7 | 1799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
