siRNA Details
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282913.1 | NP_001269842.1 | 4, 5 | 557 | ||
NM_173826.3 | NP_776187.2 | 4, 5 | 739 | |||
GenBank mRNA | AK292756.1 | - | 4, 5 | 557 | ||
AL832473.1 | - | 4, 5 | 416 | |||
BC101060.2 | - | 4, 5 | 416 | 40008144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
