siRNA Details
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282913.1 | NP_001269842.1 | 11 | 2533 | ||
NM_001282914.1 | NP_001269843.1 | 11 | 2639 | |||
NM_001282915.1 | NP_001269844.1 | 11 | 2457 | |||
NM_173826.3 | NP_776187.2 | 11 | 2715 | |||
XM_017006251.1 | XP_016861740.1 | 12 | 3667 | |||
GenBank mRNA | AK123680.1 | - | 2 | 1112 | ||
AK124087.1 | - | 9 | 1674 | |||
AL832473.1 | - | 11 | 2392 | |||
AL833118.1 | - | 10 | 2133 | |||
BC101058.2 | - | 11 | 3055 | |||
BC101059.2 | - | 11 | 2316 | 40008143 | ||
BC101060.2 | - | 11 | 2392 | 40008144 | ||
BC101061.2 | - | 11 | 2316 | 40008152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
