siRNA Details
Gene Symbol: P2RX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029075.1 | XP_016884564.1 | 1 | 15 |
Gene Symbol: THAP7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027051.1 | 2 | 1955 | |||
GenBank mRNA | AK096901.1 | - | 2 | 1660 | ||
AK097739.1 | - | 2 | 1223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
