siRNA Details
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178554.4 | NP_848649.3 | 9 | 823 | ||
XM_005247418.3 | XP_005247475.1 | 8 | 5791 | |||
XM_006713612.3 | XP_006713675.1 | 8 | 5737 | |||
XM_017006287.1 | XP_016861776.1 | 9 | 5869 | |||
XM_017006288.1 | XP_016861777.1 | 9 | 5815 | |||
XM_017006289.1 | XP_016861778.1 | 8 | 5806 | |||
XM_017006290.1 | XP_016861779.1 | 8 | 5752 | |||
GenBank mRNA | AK090526.1 | - | 9 | 823 | ||
AK126993.1 | - | 8 | 598 | |||
AK293840.1 | - | 8 | 756 | |||
AK294500.1 | - | 9 | 824 | |||
BC136349.1 | - | 9 | 784 | |||
DA212239.1 | - | 1 | 31 |
