siRNA Details
Gene Symbol: ZDHHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135179.1 | NP_001128651.1 | 7 | 10407 | ||
NM_016598.2 | NP_057682.1 | 8 | 10491 | |||
XM_005265209.1 | XP_005265266.1 | 8 | 1787 | |||
XM_005265212.2 | XP_005265269.1 | 7 | 1711 | |||
XM_006713189.1 | XP_006713252.1 | 8 | 1805 | |||
XM_006713194.3 | XP_006713257.1 | 7 | 1686 | |||
XM_011533797.1 | XP_011532099.1 | 9 | 1889 | |||
XM_011533800.2 | XP_011532102.1 | 9 | 1882 | |||
XM_017006557.1 | XP_016862046.1 | 10 | 2058 | |||
XM_017006558.1 | XP_016862047.1 | 11 | 2104 | |||
XM_017006559.1 | XP_016862048.1 | 10 | 2153 | |||
XM_017006560.1 | XP_016862049.1 | 9 | 1974 | |||
XM_017006561.1 | XP_016862050.1 | 9 | 1956 | |||
XM_017006562.1 | XP_016862051.1 | 8 | 1872 | |||
XM_017006565.1 | XP_016862054.1 | 10 | 2039 | |||
XM_017006568.1 | XP_016862057.1 | 8 | 1465 | |||
XM_017006570.1 | XP_016862059.1 | 8 | 1859 | |||
GenBank mRNA | AK127837.1 | - | 1 | 1963 | ||
AL832057.1 | - | 1 | 924 | |||
BC101284.2 | - | 1 | 1391 | |||
BC101285.2 | - | 2 | 743 | |||
BC101286.1 | - | 1 | 1391 | |||
BC101287.1 | - | 2 | 743 | |||
BC113070.1 | - | 2 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
