siRNA Details
Gene Symbol: SBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166034.1 | NP_001159506.1 | 1 | 384 | ||
NM_001166035.1 | NP_001159507.1 | 1 | 384 | |||
NM_198538.3 | NP_940940.1 | 1 | 384 | |||
XM_011526929.2 | XP_011525231.1 | 1 | 404 | |||
XM_011526930.2 | XP_011525232.1 | 1 | 360 | |||
XM_011526931.2 | XP_011525233.1 | 1 | 404 | |||
GenBank mRNA | AA055367.1 | - | 1 | 170 | ||
AK291305.1 | - | 1 | 344 | |||
AY358701.1 | - | 1 | 384 | |||
BC063640.1 | - | 1 | 326 | 4750640 | ||
BX648076.1 | - | 1 | 346 |
