siRNA Details
Gene Symbol: SORBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005775.4 | NP_005766.3 | 4 | 620 | ||
XM_006716266.1 | XP_006716329.1 | 5 | 583 | |||
XM_006716268.2 | XP_006716331.1 | 4 | 984 | |||
GenBank mRNA | AF037261.1 | - | 4 | 421 | ||
AK303950.1 | - | 4 | 431 | |||
BC067260.1 | - | 4 | 406 | 6186182 | ||
BC091514.1 | - | 4 | 620 | 6201273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
