siRNA Details
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011513.3 | NP_001011513.3 | 5, 6 | 564 | ||
NM_001256428.1 | NP_001243357.1 | 4, 5 | 739 | |||
NM_006457.4 | NP_006448.4 | 5, 6 | 891 | |||
XM_005262695.4 | XP_005262752.1 | 5, 6 | 861 | |||
XM_005262696.4 | XP_005262753.1 | 5, 6 | 479 | |||
GenBank mRNA | AF061258.1 | - | 5, 6 | 793 | ||
AK291624.1 | - | 5, 6 | 813 | |||
AK296980.1 | - | 4, 5 | 653 | |||
AL832157.1 | - | 5, 6 | 466 | |||
AY598328.1 | - | 4, 5 | 710 | |||
BC008741.2 | - | 5, 6 | 830 | 3345715 | ||
BT007328.1 | - | 4, 5 | 710 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001190855, NM_001190854, NM_001190852, NM_019808, NM_001190856, and others.
Gene Ontology Categories:
