siRNA Details
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011513.3 | NP_001011513.3 | 9 | 1065 | ||
NM_001256425.1 | NP_001243354.1 | 8 | 973 | |||
NM_001256426.1 | NP_001243355.1 | 13 | 1479 | |||
NM_001256427.1 | NP_001243356.1 | 10 | 1083 | |||
NM_001256428.1 | NP_001243357.1 | 8 | 1240 | |||
NM_006457.4 | NP_006448.4 | 9 | 1392 | |||
XM_005262693.4 | XP_005262750.1 | 13 | 1776 | |||
XM_005262695.4 | XP_005262752.1 | 12 | 1758 | |||
XM_005262696.4 | XP_005262753.1 | 12 | 1376 | |||
XM_005262698.4 | XP_005262755.1 | 10 | 1005 | |||
XM_006714066.3 | XP_006714129.1 | 14 | 1791 | |||
XM_006714068.3 | XP_006714131.1 | 14 | 1409 | |||
XM_006714069.3 | XP_006714132.1 | 11 | 1395 | |||
XM_006714070.3 | XP_006714133.1 | 14 | 1392 | |||
XM_011531543.2 | XP_011529845.1 | 13 | 1374 | |||
XM_017007657.1 | XP_016863146.1 | 10 | 1380 | |||
XM_017007658.1 | XP_016863147.1 | 11 | 1013 | |||
GenBank mRNA | AB209531.1 | - | 5 | 1104 | ||
AF061258.1 | - | 9 | 1294 | |||
AK291624.1 | - | 9 | 1314 | |||
AK296980.1 | - | 8 | 1154 | |||
AK304125.1 | - | 10 | 1005 | |||
AL832157.1 | - | 9 | 967 | |||
AL832686.1 | - | 13 | 1383 | |||
AY598328.1 | - | 8 | 1211 | |||
BC008741.2 | - | 9 | 1331 | 3345715 | ||
BC026029.2 | - | 7 | 835 | |||
BT007328.1 | - | 8 | 1211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
