siRNA Details
Gene Symbol: KIAA1429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015496.4 | NP_056311.2 | 4, 5 | 394 | ||
NM_183009.2 | NP_892121.1 | 4, 5 | 394 | |||
GenBank mRNA | AB037850.1 | - | 4, 5 | 318 | ||
AF116724.1 | - | 4, 5 | 318 | |||
AL832487.1 | - | 4, 5 | 337 | |||
BC112288.1 | - | 4, 5 | 406 | 8327756 | ||
BC113380.1 | - | 4, 5 | 394 | 8322432 |
