siRNA Details
Gene Symbol: WWC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015691.3 | NP_056506.2 | 3 | 272 | ||
GenBank mRNA | AI306162.1 | - | 4 | 336 | ||
AK292961.1 | - | 2 | 154 | |||
BU500300.1 | - | 3 | 480 | |||
BU500742.1 | - | 3 | 456 | |||
KC987947.1 | - | 3 | 449 |
