siRNA Details
Gene Symbol: FAM198B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128424.1 | NP_001121896.1 | 3 | 1373 | ||
NM_016613.6 | NP_057697.2 | 3 | 1418 | |||
GenBank mRNA | AF223468.1 | - | 2 | 360 | ||
AK304112.1 | - | 2 | 298 | |||
AK315768.1 | - | 2 | 489 | |||
AL832589.1 | - | 3 | 1244 | |||
AL834177.1 | - | 3 | 869 | |||
AY358785.1 | - | 3 | 1335 | |||
BC017701.1 | - | 2 | 321 | |||
BC100016.1 | - | 2 | 257 | |||
CR457196.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
