siRNA Details
Gene Symbol: PLXNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017514.4 | NP_059984.3 | 32 | 5667 | ||
XM_005274705.4 | XP_005274762.2 | 32 | 5607 | |||
XM_005274706.4 | XP_005274763.2 | 32 | 5339 | |||
XM_006724829.3 | XP_006724892.1 | 32 | 5571 | |||
XM_011531183.2 | XP_011529485.1 | 31 | 5031 | |||
GenBank mRNA | X87852.1 | - | 32 | 5676 |
