siRNA Details
Gene Symbol: MARCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166373.1 | NP_001159845.1 | 8 | 2084 | ||
NM_017923.3 | NP_060393.1 | 4 | 1971 | |||
XM_011532055.2 | XP_011530357.1 | 8 | 2789 | |||
XM_017008334.1 | XP_016863823.1 | 7 | 2011 | |||
XM_017008335.1 | XP_016863824.1 | 8 | 2390 | |||
XM_017008336.1 | XP_016863825.1 | 8 | 2256 | |||
GenBank mRNA | AK000675.1 | - | 4 | 1864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
