siRNA Details
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018013.3 | NP_060483.3 | 7 | 3614 | ||
XM_005267041.4 | XP_005267098.1 | 8 | 3775 | |||
XM_005267042.4 | XP_005267099.1 | 7 | 3680 | |||
XM_011535921.2 | XP_011534223.1 | 8 | 3299 | |||
XM_011535923.2 | XP_011534225.1 | 3 | 2513 | |||
XM_017010991.1 | XP_016866480.1 | 6 | 4137 | |||
XM_017010992.1 | XP_016866481.1 | 6 | 2906 | |||
XM_017010993.1 | XP_016866482.1 | 6 | 2936 | |||
XM_017010994.1 | XP_016866483.1 | 6 | 2895 |
