siRNA Details
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003141.3 | NP_003132.2 | 2 | 232 | ||
GenBank mRNA | AK225564.1 | - | 2 | 215 | ||
AY742713.1 | - | 1 | 119 | |||
BC010861.1 | - | 2 | 203 | 3900540 | ||
M34551.1 | - | 1 | 157 | |||
M62800.1 | - | 2 | 204 |
