siRNA Details
Gene Symbol: RC3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100588.1 | NP_001094058.1 | 9 | 1528 | ||
NM_018835.2 | NP_061323.2 | 9 | 1528 | |||
GenBank mRNA | AF255303.1 | - | 8 | 1888 | ||
AK000308.1 | - | 7 | 962 | |||
AK226010.1 | - | 7 | 962 | |||
AL833177.1 | - | 9 | 1467 | |||
AL834431.1 | - | 3 | 279 | |||
BC044642.1 | - | 9 | 1460 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001100591, NM_001290642, BC117952, AK053071, BC117951.
Gene Ontology Categories:
