siRNA Details
Gene Symbol: ACTR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040135.2 | NP_001035225.1 | 9 | 1009 | ||
NM_020445.5 | NP_065178.1 | 9 | 1009 | |||
NR_073000.1 | 8 | 953 | ||||
NR_073001.1 | 8 | 884 | ||||
XM_006716065.2 | XP_006716128.1 | 9 | 991 | |||
XM_006716066.1 | XP_006716129.1 | 9 | 991 | |||
XM_006716067.3 | XP_006716130.1 | 9 | 991 | |||
XM_011516425.2 | XP_011514727.1 | 9 | 991 | |||
XM_011516426.2 | XP_011514728.1 | 9 | 991 | |||
XM_011516427.1 | XP_011514729.1 | 9 | 991 | |||
XM_017012460.1 | XP_016867949.1 | 9 | 991 | |||
XM_017012461.1 | XP_016867950.1 | 9 | 991 | |||
XM_017012462.1 | XP_016867951.1 | 9 | 920 | |||
XM_017012463.1 | XP_016867952.1 | 9 | 859 | |||
XM_017012464.1 | XP_016867953.1 | 8 | 950 | |||
XM_017012465.1 | XP_016867954.1 | 8 | 813 | |||
GenBank mRNA | AB209174.1 | - | 5 | 4336 | ||
AF023453.1 | - | 9 | 911 | |||
AF086920.1 | - | 8 | 875 | |||
AK001198.1 | - | 5 | 477 | |||
AK294292.1 | - | 8 | 907 | |||
AK315534.1 | - | 9 | 991 | |||
AK316078.1 | - | 8 | 804 | |||
BC008682.1 | - | 9 | 964 | 3854008 | ||
BC015207.1 | - | 9 | 921 | 3916310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
