siRNA Details
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 4 | 521 | ||
NM_001170570.1 | NP_001164041.1 | 3 | 397 | |||
NM_022101.3 | NP_071384.1 | 4 | 439 | |||
GenBank mRNA | AK096297.1 | - | 4 | 485 | ||
AK298877.1 | - | 3 | 375 | |||
BC023506.2 | - | 4 | 400 | |||
BQ223940.1 | - | 4 | 404 | |||
DA590933.1 | - | 4 | 523 | |||
DA921725.1 | - | 4 | 414 | |||
DB033119.1 | - | 4 | 454 |
